5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

062537-500MG
101924-060EA 484.62 USD
101924-060
5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid
5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆Cl₂N₂O₂
MW: 257.0700 g/mol
MDL Number: MFCD03030206
CAS Number: 276684-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR