Methyl 3,6-dichlorobenzo[b]thiophene-2-carboxylate

Supplier: Matrix Scientific

062530-500MG
101924-046EA 309.92 USD
101924-046
Methyl 3,6-dichlorobenzo[b]thiophene-2-carboxylate
Methyl 3,6-dichlorobenzo[b]thiophene-2-carboxylate

CAS: 21211-18-7
MDL Number: MFCD00781759
Molecular Formula: C10H6Cl2O2S
Molecular Weight: 261,13
Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR