1-(3-Chloropropanoyl)-3,5-dimethylpiperidine

Supplier: Matrix Scientific

062464-500MG
101923-976EA 183.08 USD
101923-976
1-(3-Chloropropanoyl)-3,5-dimethylpiperidine
1-(3-Chloropropanoyl)-3,5-dimethylpiperidine

Hazard: Irritant

Formula: C₁₀H₁₈ClNO
CAS Number: 349097-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR