4-sec-Butylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-sec-Butylbenzeneboronic acid

062411-500MG
101923-908EA 158.46 USD
101923-908
4-sec-Butylphenylboronic acid
4-sec-Butylphenylboronic acid

Hazard: Irritant

Formula: C₁₀H₁₅BO₂
MW: 178.04
MDL Number: MFCD02179456
CAS Number: 850568-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR