2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde

Supplier: Matrix Scientific

062410-500MG
101923-906EA 364.62 USD
101923-906
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀N₂O
MW: 186.2099 g/mol
MDL Number: MFCD08059904
CAS Number: 1015845-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR