2,3-Dihydrobenzo[b]furan-7-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydrobenzofuran-7-carboxylic acid

062409-500MG
101923-904EA 183.08 USD
101923-904
2,3-Dihydrobenzo[b]furan-7-carboxylic acid
2,3-Dihydrobenzo[b]furan-7-carboxylic acid

Hazard: Irritant

Formula: C₉H₈O₃
MW: 164.1580 g/mol
MDL Number: MFCD00671526
CAS Number: 35700-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR