Sulfachloropyridazine

Supplier: Matrix Scientific

Synonyms: 4-Amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide4-Amino-N-(6-chloropyridazin-3-yl)benzenesulphonamide

062264-500MG
101923-620EA 243.08 USD
101923-620
Sulfachloropyridazine
Sulphachloropyridazine

Hazard: Irritant

Formula: C₁₀H₉ClN₄O₂S
MW: 284.7221 g/mol
MDL Number: MFCD00057371
CAS Number: 80-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR