4-(2-Phenylethoxy)benzoyl chloride

Supplier: Matrix Scientific

062161-500MG
101923-414EA 221.54 USD
101923-414
4-(2-Phenylethoxy)benzoyl chloride
4-(2-Phenylethoxy)benzoyl chloride

Hazard: Irritant

Formula: C₁₅H₁₃ClO₂
MDL Number: MFCD12198033
CAS Number: 756478-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR