2-Methyl-2-(3-methyl-4-nitrophenoxy)propanoyl chloride

Supplier: Matrix Scientific

061915-500MG
101922-922EA 243.08 USD
101922-922
2-Methyl-2-(3-methyl-4-nitrophenoxy)propanoyl chloride
2-Methyl-2-(3-methyl-4-nitrophenoxy)propanoyl chloride

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₄
MDL Number: MFCD12197857
CAS Number: 1160257-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR