2-(4-Fluorophenoxy)propanoyl chloride

Supplier: Matrix Scientific

061480-500MG
101922-052EA 364.62 USD
101922-052
2-(4-Fluorophenoxy)propanoyl chloride
2-(4-Fluorophenoxy)propanoyl chloride

Hazard: Irritant

Formula: C₉H₈ClFO₂
MDL Number: MFCD02295758
CAS Number: 56895-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR