(2-Fluorophenoxy)acetyl chloride

Supplier: Matrix Scientific

061479-500MG
101922-050EA 243.08 USD
101922-050
(2-Fluorophenoxy)acetyl chloride
(2-Fluorophenoxy)acetyl chloride

Hazard: Irritant

Formula: C₈H₆ClFO₂
MDL Number: MFCD02295747
CAS Number: 2965-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR