5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonyl chloride

Supplier: Matrix Scientific

061476-500MG
101922-044EA 606.15 USD
101922-044
5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonyl chloride
5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonyl chloride

Hazard: Irritant

Formula: C₁₄H₆BrClF₃N₃O
MW: 404.58 g/mol
MDL Number: MFCD12197769
CAS Number: 848422-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR