(4-Nitrophenoxy)acetyl chloride

Supplier: Matrix Scientific

061474-500MG
101922-040EA 336.92 USD
101922-040
(4-Nitrophenoxy)acetyl chloride
(4-Nitrophenoxy)acetyl chloride

Hazard: Irritant

Formula: C₈H₆ClNO₄
MDL Number: MFCD01027039
CAS Number: 20142-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR