3,5-Dimethyladamantane-1-carbonyl chloride

Supplier: Matrix Scientific

061470-500MG
101922-032EA 364.62 USD
101922-032
3,5-Dimethyladamantane-1-carbonyl chloride
3,5-Dimethyladamantane-1-carbonyl chloride

Hazard: Irritant

Formula: C₁₃H₁₉ClO
MDL Number: MFCD12197768
CAS Number: 3124-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR