1-Phenyl-1H-pyrazole-4-carbonyl chloride

Supplier: Matrix Scientific

061469-500MG
101922-030EA 404.62 USD
101922-030
1-Phenyl-1H-pyrazole-4-carbonyl chloride
1-Phenyl-1H-pyrazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₀H₇ClN₂O
MDL Number: MFCD11169644
CAS Number: 267641-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR