2-(4-Methoxyphenyl)-6-methylquinoline-4-carbonyl chloride

Supplier: Matrix Scientific

061467-500MG
101922-026EA 364.62 USD
101922-026
2-(4-Methoxyphenyl)-6-methylquinoline-4-carbonyl chloride
2-(4-Methoxyphenyl)-6-methylquinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₈H₁₄ClNO₂
MDL Number: MFCD03421158
CAS Number: 1160264-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR