2-(4-Methylphenyl)quinoline-4-carbonyl chloride

Supplier: Matrix Scientific

061466-500MG
101922-024EA 484.62 USD
101922-024
2-(4-Methylphenyl)quinoline-4-carbonyl chloride
2-(4-Methylphenyl)quinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₇H₁₂ClNO
MDL Number: MFCD03421045
CAS Number: 860112-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR