2-(4-Ethylphenyl)quinoline-4-carbonyl chloride

Supplier: Matrix Scientific

061465-500MG
101922-022EA 364.62 USD
101922-022
2-(4-Ethylphenyl)quinoline-4-carbonyl chloride
2-(4-Ethylphenyl)quinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₈H₁₄ClNO
MDL Number: MFCD03421051
CAS Number: 1160264-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR