3-Chlorobenzo[b]thiophene-2-carbonyl chlorid

Supplier: Matrix Scientific

Synonyms: 3-Chlorobenzo[b]thiophene-2-carbonyl chloride

061458-500MG
101922-006EA 146.15 USD
101922-006
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid

Hazard: Irritant

Formula: C₉H₄Cl₂OS
MW: 231.10 g/mol
MDL Number: MFCD00053069
CAS Number: 21815-91-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR