N-(4-Aminobutyl)-N-ethylisoluminol
Supplier: Matrix Scientific
Synonyms:
6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione
, ABEI
061452-500MG
101921-994EA
1818.46
USD
101921-994
N-(4-Aminobutyl)-N-ethylisoluminol
N-(4-Aminobutyl)-N-ethylisoluminol
CAS: 66612-29-1
MDL Number: MFCD00010559
Molecular Formula: C14H20N4O2
Molecular Weight: 276,34
Hazard: Irritant
Formula:
C₁₄H₂₀N₄O₂ MW: 276.34 g/mol |
MDL Number:
MFCD00010559 CAS Number: 66612-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...