5,6-Dihydrofuro[2,3-d]pyrimidin-2-amine

Supplier: Matrix Scientific

061451-500MG
101921-992EA 606.15 USD
101921-992
5,6-Dihydrofuro[2,3-d]pyrimidin-2-amine
5,6-Dihydrofuro[2,3-d]pyrimidin-2-amine

Hazard: Irritant

Formula: C₆H₇N₃O
MDL Number: MFCD12197763
CAS Number: 88513-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR