2-Amino-5-(2-chloroethyl)pyrimidin-4-ol

Supplier: Matrix Scientific

061450-500MG
101921-990EA 484.62 USD
101921-990
2-Amino-5-(2-chloroethyl)pyrimidin-4-ol
2-Amino-5-(2-chloroethyl)pyrimidin-4-ol

Hazard: Irritant

Formula: C₆H₈ClN₃O
MDL Number: MFCD12197762
CAS Number: 805180-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR