(1-Methyl-1H-pyrrol-2-yl)(oxo)acetic acid

Supplier: Matrix Scientific

061447-500MG
101921-984EA 484.62 USD
101921-984
(1-Methyl-1H-pyrrol-2-yl)(oxo)acetic acid
(1-Methyl-1H-pyrrol-2-yl)(oxo)acetic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MDL Number: MFCD11983457
CAS Number: 21898-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR