[2-(Tetrahydro-2H-pyran-3-yl)ethyl]amine

Supplier: Matrix Scientific

061444-500MG
101921-980EA 606.15 USD
101921-980
[2-(Tetrahydro-2H-pyran-3-yl)ethyl]amine
[2-(Tetrahydro-2H-pyran-3-yl)ethyl]amine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD08234897
CAS Number: 98430-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR