2-Methyltetrahydro-2H-pyran-2-carboxylic acid

Supplier: Matrix Scientific

061443-500MG
101921-978EA 606.15 USD
101921-978
2-Methyltetrahydro-2H-pyran-2-carboxylic acid
2-Methyltetrahydro-2H-pyran-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₂O₃
MDL Number: MFCD09864362
CAS Number: 4180-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR