Pyridine-2,5-dicarboxamide

Supplier: Matrix Scientific

061441-500MG
101921-974EA 167.69 USD
101921-974
Pyridine-2,5-dicarboxamide
Pyridine-2,5-dicarboxamide

Hazard: Irritant

Formula: C₇H₇N₃O₂
MDL Number: MFCD00457415
CAS Number: 4663-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR