5-[2-Chloro-3-(trifluoromethyl)phenoxy]-2-furoic acid

Supplier: Matrix Scientific

061439-500MG
101921-970EA 243.08 USD
101921-970
5-[2-Chloro-3-(trifluoromethyl)phenoxy]-2-furoic acid
5-[2-Chloro-3-(trifluoromethyl)phenoxy]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₆ClF₃O₄
MDL Number: MFCD12197756
CAS Number: 1160264-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR