2-[2-Fluoro-3-(trifluoromethyl)phenoxy]isonicotinic acid
Supplier: Matrix Scientific
061437-500MG
101921-966EA
243.08
USD
101921-966
2-[2-Fluoro-3-(trifluoromethyl)phenoxy]isonicotinic acid
2-[2-Fluoro-3-(trifluoromethyl)phenoxy]isonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₇F₄NO₃ |
MDL Number:
MFCD12197753 CAS Number: 1160264-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...