6-(2,3-Dihydro-1H-inden-5-yloxy)nicotinic acid

Supplier: Matrix Scientific

061436-500MG
101921-964EA 243.08 USD
101921-964
6-(2,3-Dihydro-1H-inden-5-yloxy)nicotinic acid
6-(2,3-Dihydro-1H-inden-5-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₃
MDL Number: MFCD12197752
CAS Number: 1042778-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR