1,2,3,4-Tetrahydroquinoline-2-carboxamide

Supplier: Matrix Scientific

061434-500MG
101921-960EA 243.08 USD
101921-960
1,2,3,4-Tetrahydroquinoline-2-carboxamide
1,2,3,4-Tetrahydroquinoline-2-carboxamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD09811961
CAS Number: 91842-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR