N-[2-(4-Ethylpiperazin-1-yl)-1-methyl-2-oxoethyl]-2,2,2-trifluoroacetamide

Supplier: Matrix Scientific

061433-500MG
101921-958EA 146.15 USD
101921-958
N-[2-(4-Ethylpiperazin-1-yl)-1-methyl-2-oxoethyl]-2,2,2-trifluoroacetamide
N-[2-(4-Ethylpiperazin-1-yl)-1-methyl-2-oxoethyl]-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₁₁H₁₈F₃N₃O₂
MDL Number: MFCD12197748
CAS Number: 1214235-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR