N-Phenyl-2-[(pyridin-2-ylmethyl)amino]ethanesulfonamide
Supplier: Matrix Scientific
061432-500MG
101921-956EA
364.62
USD
101921-956
N-Phenyl-2-[(pyridin-2-ylmethyl)amino]ethanesulfonamide
N-Phenyl-2-[(pyridin-2-ylmethyl)amino]ethanesulfonamide
Hazard: Irritant
Formula:
C₁₄H₁₇N₃O₂S MW: 291.37 g/mol |
MDL Number:
MFCD12197747 CAS Number: 1160264-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...