(2Z)-3,5-Diamino-2-cyano-5-oxopent-2-enoic acid

Supplier: Matrix Scientific

061429-500MG
101921-950EA 193.85 USD
101921-950
(2Z)-3,5-Diamino-2-cyano-5-oxopent-2-enoic acid
(2Z)-3,5-Diamino-2-cyano-5-oxopent-2-enoic acid

Hazard: Irritant

Formula: C₆H₇N₃O₃
MDL Number: MFCD12197742
CAS Number: 1160264-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR