Isopropoxyacetic acid

Supplier: Matrix Scientific

061424-500MG
101921-940EA 115.38 USD
101921-940
Isopropoxyacetic acid
Isopropoxyacetic acid

Hazard: Irritant

Formula: C₅H₁₀O₃
Boiling Pt: 113 °C
MDL Number: MFCD09055367
CAS Number: 33445-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR