5-Chloro-N-(2-pyrrolidin-1-ylethyl)pyrazine-2-carboxamide

Supplier: Matrix Scientific

061419-500MG
101921-930EA 484.62 USD
101921-930
5-Chloro-N-(2-pyrrolidin-1-ylethyl)pyrazine-2-carboxamide
5-Chloro-N-(2-pyrrolidin-1-ylethyl)pyrazine-2-carboxamide

Hazard: Irritant

Formula: C₁₁H₁₅ClN₄O
MDL Number: MFCD12197736
CAS Number: 1160264-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR