1-(5-Carboxy-4-methyl-1,3-thiazol-2-yl)-1H-indole-4-carboxylic acid
Supplier: Matrix Scientific
061417-500MG
101921-926EA
243.08
USD
101921-926
1-(5-Carboxy-4-methyl-1,3-thiazol-2-yl)-1H-indole-4-carboxylic acid
1-(5-Carboxy-4-methyl-1,3-thiazol-2-yl)-1H-indole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₀N₂O₄S |
MDL Number:
MFCD12197735 CAS Number: 1160264-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...