2-(3,4-Dimethoxyphenyl)indane-2-carbonitrile

Supplier: Matrix Scientific

061410-500MG
101921-912EA 606.15 USD
101921-912
2-(3,4-Dimethoxyphenyl)indane-2-carbonitrile
2-(3,4-Dimethoxyphenyl)indane-2-carbonitrile

Hazard: Irritant

Formula: C₁₈H₁₇NO₂
MDL Number: MFCD00565264
CAS Number: 1160264-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR