5-Propyl-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

061409-500MG
101921-910EA 484.62 USD
101921-910
5-Propyl-1,3,4-oxadiazol-2-amine
5-Propyl-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₅H₉N₃O
MDL Number: MFCD09261278
CAS Number: 69741-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR