(1-Cyclopentylpyrrolidin-3-yl)methanol

Supplier: Matrix Scientific

061406-500MG
101921-906EA 606.15 USD
101921-906
(1-Cyclopentylpyrrolidin-3-yl)methanol
(1-Cyclopentylpyrrolidin-3-yl)methanol

Hazard: Irritant

Formula: C₁₀H₁₉NO
MDL Number: MFCD10003884
CAS Number: 1017429-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR