(1-tert-Butylpyrrolidin-3-yl)methanol

Supplier: Matrix Scientific

061404-500MG
101921-902EA 606.15 USD
101921-902
(1-tert-Butylpyrrolidin-3-yl)methanol
(1-tert-Butylpyrrolidin-3-yl)methanol

Hazard: Irritant

Formula: C₉H₁₉NO
MDL Number: MFCD10003916
CAS Number: 71548-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR