(2-Morpholin-4-ylbutyl)amine

Supplier: Matrix Scientific

061402-500MG
101921-898EA 484.62 USD
101921-898
(2-Morpholin-4-ylbutyl)amine
(2-Morpholin-4-ylbutyl)amine

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD09727564
CAS Number: 953888-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR