[2-(4-Methylpiperazin-1-yl)butyl]amine

Supplier: Matrix Scientific

061401-500MG
101921-896EA 484.62 USD
101921-896
[2-(4-Methylpiperazin-1-yl)butyl]amine
[2-(4-Methylpiperazin-1-yl)butyl]amine

Hazard: Irritant

Formula: C₉H₂₁N₃
MDL Number: MFCD09729869
CAS Number: 953739-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR