2-(4-Aminophenyl)-6-bromo-8-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

061394-500MG
101921-882EA 364.62 USD
101921-882
2-(4-Aminophenyl)-6-bromo-8-methylquinoline-4-carboxylic acid
2-(4-Aminophenyl)-6-bromo-8-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₃BrN₂O₂
MDL Number: MFCD12197727
CAS Number: 1160264-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR