4-(5,6-Dichloro-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Supplier: Matrix Scientific

061393-500MG
101921-880EA 364.62 USD
101921-880
4-(5,6-Dichloro-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid
4-(5,6-Dichloro-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₃Cl₂NO₄
MDL Number: MFCD06147887
CAS Number: 1005057-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR