2-(3-Aminophenyl)-6-bromoquinoline-4-carboxylic acid

Supplier: Matrix Scientific

061388-500MG
101921-870EA 388.7 USD
101921-870
2-(3-Aminophenyl)-6-bromoquinoline-4-carboxylic acid
2-(3-Aminophenyl)-6-bromoquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₁BrN₂O₂
MDL Number: MFCD02055898
CAS Number: 438531-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR