2-Methyl-5-(4-oxo-3,4,5,6,7,8-hexahydrophthalazin-1-yl)benzenesulfonyl chloride

Supplier: Matrix Scientific

061386-500MG
101921-866EA 364.62 USD
101921-866
2-Methyl-5-(4-oxo-3,4,5,6,7,8-hexahydrophthalazin-1-yl)benzenesulfonyl chloride
2-Methyl-5-(4-oxo-3,4,5,6,7,8-hexahydrophthalazin-1-yl)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₃S
MDL Number: MFCD12197721
CAS Number: 1160264-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR