1,3-Dioxo-2-[3-(trifluoromethyl)phenyl]isoindoline-5-carboxylic acid

Supplier: Matrix Scientific

061384-500MG
101921-862EA 364.62 USD
101921-862
1,3-Dioxo-2-[3-(trifluoromethyl)phenyl]isoindoline-5-carboxylic acid
1,3-Dioxo-2-[3-(trifluoromethyl)phenyl]isoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₈F₃NO₄
MDL Number: MFCD01174923
CAS Number: 328555-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR