1-[4-(3,5-Dimethylphenoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

061379-500MG
101921-852EA 364.62 USD
101921-852
1-[4-(3,5-Dimethylphenoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid
1-[4-(3,5-Dimethylphenoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₉NO₄
MDL Number: MFCD12197720
CAS Number: 1160264-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR