2-(2,6-Diethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
Supplier: Matrix Scientific
061377-500MG
101921-848EA
243.08
USD
101921-848
2-(2,6-Diethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(2,6-Diethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₉H₁₇NO₄ |
MDL Number:
MFCD04113793 CAS Number: 100844-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...