2-Bromo-N-(4-bromo-2,3-dimethylphenyl)acetamide

Supplier: Matrix Scientific

061376-500MG
101921-846EA 364.62 USD
101921-846
2-Bromo-N-(4-bromo-2,3-dimethylphenyl)acetamide
2-Bromo-N-(4-bromo-2,3-dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₁Br₂NO
MDL Number: MFCD06149134
CAS Number: 895854-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR